1-Bromo-3-chloro-2-fluorobenzene 95%

Supplier: Thermo Scientific Chemicals
Warning

H61622-14 H61622-22
CAAAH61622-14EA 83.27 CAD
CAAAH61622-14 CAAAH61622-22
1-Bromo-3-chloro-2-fluorobenzene 95%
1-Bromo-3-chloro-2-fluorobenzene

1-Bromo-3-chloro-2-fluorobenzene, 95%

Formula: C₆H₃BrClF
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03412182
CAS Number: 144584-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR