7-Nitro-1,2,3,4-tetrahydroquinoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2,3,4-Tetrahydro-7-nitroquinoline
H61612-03
H61612-06
CAAAH61612-03EA
72.05
CAD
CAAAH61612-03
CAAAH61612-06
7-Nitro-1,2,3,4-tetrahydroquinoline 95%
7-Nitro-1,2,3,4-tetrahydroquinoline
7-Nitro-1,2,3,4-tetrahydroquinoline, 95%
Purity: 95%
CAS No.: 30450-62-5
Molecular Formula: C9H10N2O2
Formula Weight: 178.19
Physical Form: Powder
Appearance: Red
MDL No.: MFCD00496654
Formula:
C₉H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00496654 CAS Number: 30450-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...