Ethyl-2-aminothiophene-3-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-aminothiophene-3-carboxylate

H61559-14 H61559-22
CAAAH61559-14EA 189.6 CAD
CAAAH61559-14 CAAAH61559-22
Ethyl-2-aminothiophene-3-carboxylate 98%
Ethyl 2-aminothiophene-3-carboxylate

Ethyl 2-aminothiophene-3-carboxylate, 98%

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01566303
CAS Number: 31891-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR