4-Fluoro-3-nitroanisole 95%

Supplier: Thermo Scientific Chemicals
Warning

H61554-03 H61554-06
CAAAH61554-03EA 219.62 CAD
CAAAH61554-03 CAAAH61554-06 CAAAH61554-MD
4-Fluoro-3-nitroanisole 95%
4-Fluoro-3-nitroanisole

4-Fluoro-3-nitroanisole, 95%

Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD11848442
CAS Number: 61324-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR