3-Bromo-5-fluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61544-14
H61544-22
CAAAH61544-14EA
141.09
CAD
CAAAH61544-14
CAAAH61544-22
3-Bromo-5-fluorotoluene 98%
3-Bromo-5-fluorotoluene
3-Bromo-5-fluorotoluene, 98%
Formula:
C₇H₆BrF Storage Temperature: Ambient |
MDL Number:
MFCD01861195 CAS Number: 202865-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...