Methyl-4-fluoro-3-nitrobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-fluoro-3-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

H61542-14 H61542-22
CAAAH61542-14EA 219.62 CAD
CAAAH61542-14 CAAAH61542-22
Methyl-4-fluoro-3-nitrobenzoate 98%
Methyl 4-fluoro-3-nitrobenzoate

Methyl 4-fluoro-3-nitrobenzoate, 98%

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Boiling Pt: 109 °C/1 mm
Melting Pt: 56…59 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00196161
CAS Number: 329-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR