3-Maleimidopropionic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine
H61536-03
H61536-06
CAAAH61536-03EA
265.44
CAD
CAAAH61536-03
CAAAH61536-06
3-Maleimidopropionic acid 95%
3-Maleimidopropionic acid
3-Maleimidopropionic acid, 95%
Purity: 95%
CAS No.: 7423-55-4
Molecular Formula: C7H7NO4
Formula Weight: 169.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 108-112°
Beilstein Reference: 1528952
MDL No.: MFCD00043030
Formula:
C₇H₇NO₄ Melting Pt: 108…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043030 CAS Number: 7423-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...