(2-Fluorophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals
Warning

H61535-06 H61535-14
CAAAH61535-06EA 103.65 CAD
CAAAH61535-06 CAAAH61535-14
(2-Fluorophenyl)acetone 98%
(2-Fluorophenyl)acetone

2-Fluorophenylacetone, 98%

Formula: C₉H₉FO
Boiling Pt: 47 °C
Density: 1.077
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000325
CAS Number: 2836-82-0
EINECS: 220-627-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR