Polyshield® Fluid Barrier, Ahlstrom
Supplier: Ahlstrom
4002-4650 4002-4657 4002-9250
47745-880EA 497.29 CAD
47745-880 47745-882 47745-884
Polyshield® Fluid Barrier, Ahlstrom
Floor Mats Tacky Mats
Ahlstrom's Polyshield® Fluid barrier is a super absorbent, pure cellulose paper with an impermeable polyethylene backing that absorbs splatters and spills, and protects work areas from contamination and damage by toxic, infectious, corrosive, radioactive or staining materials.
Polyshield's smooth, firm surface does not inhibit stability of bottles or tubes and allows recovery of dry spills. Liquid samples can be recovered by washing the absorbed spill from the inert surface with the appropriate buffer. Disposal is safe and easy and Polyshield ashes readily upon incineration.