Stomacher® Bags for Stomacher® 3500 Lab Blender Series, Seward
Supplier: Seward Laboratory System
030020316
58976-945PK 657.61 CAD
58976-945
Stomacher® Bags for Stomacher® 3500 Lab Blender Series, Seward
Bags Blender Bags
The Stomacher® Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.
  • Made from high quality food grade virgin LDPE granule extruded to 65 µm film thickness
  • Withstand extensive blending without tears or perforation
  • Low cost option for sample preparation with no risk of cross contamination between samples
  • Irradiated sterile
Bags for the Stomacher® 3500 have a capacity of 400 to 3500 ml. Compatible with all lab blenders and homogenizers.
Ordering information: Irradiation certificates are available on request.