SP Bel-Art Circulus™ Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37170-0000 F37172-0000
30617-696CS 1589.65 CAD
30617-696 30617-698
SP Bel-Art Circulus™ Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Circulus™ Magnetic Stirring Bars offer fast mixing action and strong turbulence as well as greater stability with less drifting at higher speeds.
  • Ideal for containers with round or convex bottoms
  • Fast mixing action
  • Solid PTFE end disks
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.