SP Bel-Art Spinstar® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37146-0250 F37146-0400 F37146-0100
70680-164EA 130.38 CAD
70680-164 70680-166 70680-226
SP Bel-Art Spinstar® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Spinstar® magnetic stirring bars create a deep mixing vortex at relatively slow speeds.
  • Perfect for applications requiring slow, thorough mixing
  • PTFE low-friction coating
  • Durable and inert
  • Autoclavable
Star shape is designed to fit the inside diameter of most commonly-used beakers.
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.