SP Bel-Art Spinring® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37140-0010 F37140-0020
47750-214EA 87.17 CAD
47750-214 74950-304 74950-306 76006-480
SP Bel-Art Spinring® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Spinring® stirring bars provide maximum stabilization of the magnetic stirring bar with the addition of a removable “hoop” around a standard octagonal bar.
  • Best suited for larger open-neck vessels, such as buckets and beakers
  • Friction fit of the “hoop” and bar allows them to spin as a unit
  • "Spin out” is virtually eliminated
  • Stirring bar remains centered in vessel
  • Octagon shape creates a deep vortex without splashing or chattering
  • PTFE low-friction coating
  • Durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.