Puritan® Uni Tranz-RT Transport System, Media Only, Puritan Medical Products
Supplier: Puritan Medical Products
UT-100
UT-300
89233-456CS
747.51
CAD
89233-456
89233-458
Puritan® Uni Tranz-RT Transport System, Media Only, Puritan Medical Products
Media for Microbiology
Prepared Media for Microbiology
Puritan® Uni Tranz-RT Transport Systems are used for the collection and preservation of Virus - Chlamydia - Mycoplasma and Ureaplasma
Uni Tranz-RT Transport Systems are Universal Transport Media, stable at room temperature. These systems include media only. Swabs are not included.
Ordering information: Products are final sale.