DNAmark™ DNA Plus Ladders, G-Biosciences
Supplier: G-Biosciences
786-854 786-855 786-856
CA89233-910EA 305.38 CAD
CA89233-910 CA89233-912 CA89233-914
DNAmark™ DNA Plus Ladders, G-Biosciences
Electrophoresis Reagents Molecular Weight Markers
DNAmark™ 1kb Plus contains 15 DNA bands ranging from 100bp to 10Kb
DNAmark™ 100bp Ladder consists of 11 fragments ranging from 100-1500 base pairs (bp). The 500bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5µl loading, all fragments except 500bp are 40ng. The 500bp fragment is 100ng. This ladder is pre-mixed with loading dye and is ready to use.
DNAmark™ 100bp Plus Ladder consists of 14 DNA fragments ranging in size from 100-3000 base pairs (bp). The 500bp and 1200bp fragment are present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500bp and 1200bp are 40ng. The 500bp and 1200bp fragment are 100ng. This ladder is pre-mixed with loading dye and is ready to use.
The 500bp and 3,000bp fragment are present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5µl loading, all fragments except 500bp and 3,000bp are 40ng. The 500bp and 3,000bp fragment are 100ng. This ladder is pre-mixed with loading dye and is ready to use.