CryoSavers™ Cryo Preservation System
Supplier: HARDY DIAGNOSTICS CA
CS100B CS81BNB CS100BNB CSM100 CS81B
89501-502EA 917.46 CAD
89501-502 CA52428-856 CA89423-726 CA10001-416 CA10063-768
CryoSavers™ Cryo Preservation System
Cryopreservation Systems
The CryoSaver system is designed for ease of use
  • Convenient, ready-to-use media saves you time and expense
  • Media formulations allow for both the growth and long-term preservation of the organism
  • Glycerol is used as a cryoprotectant
  • An advanced "honey-comb" box design (with numbered compartments) protects your CryoSaver during shipping and saves valuable freezer space in the lab
  • Ergonomically designed caps feature ridges that allow for easy opening
  • Threads are located on the outside of the vial to reduce the chance of contamination
  • Each vial has a write-on
More Product Information