SP Bel-Art Electrode Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H18509-0000
34102-089EA 244.93 CAD
34102-089
SP Bel-Art Electrode Rack, Bel-Art Products, a part of SP
Electrode Holders
Handy rack holds 8 test tubes for storing and conditioning pH and ion electrodes.
  • Polypropylene construction
  • Suitable for storing and conditioning electrodes
  • Chemically resistant
  • Autoclavable
The polypropylene rack resists chemical attack and is autoclavable at 121 °C (250 °F). Rack accomodates two rows of 4 tubes, up to 20 mm in diameter.
LxWxH: 10.8 x 6 x 12.1 cm