Shell Vials, 1 ml, Ace Glass
Supplier: Ace Glass
5698-12 5698-14 5698-20 5698-24
89427-710PK 106.88 CAD
89427-710 89427-712 10767-242 10767-244
Shell Vials, 1 ml, Ace Glass
Vials
Vials are made from type 1 borosilicate glass with snap plug cap.
Available in clear or amber.