SP Bel-Art Spinplus® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37144-0112 F37144-0114 F37144-0100 F37144-0034 F37144-0038
70690-166EA 43.21 CAD
70690-166 70690-168 70690-170 70690-172 70690-174
SP Bel-Art Spinplus® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • PTFE low-friction coating
  • Durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.