SP Bel-Art Spinbar® Disposable Polygon Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37112-0001 F37112-0012 F37112-0016 F37112-0002
30617-682EA 189.97 CAD
30617-682 30617-684 30617-686 76001-832
SP Bel-Art Spinbar® Disposable Polygon Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single-use applications mixing radioactive reagents or when cross-contamination might be a concern
  • PTFE low-friction coating
  • Durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.