CryoPlus AutoFill Cryogenic Storage Systems, Thermo Scientific
Supplier: Thermo Scientific
7400 7402 7404 7406 7400 7404 7402
CA89510-258EA 51034 CAD
CA89510-258 CA89510-254 89510-256 CA89510-256 89510-258 89510-254 89510-260
CryoPlus AutoFill Cryogenic Storage Systems, Thermo Scientific
Cryopreservation Systems
Our CryoPlus LN2 storage systems provide precise and accurate control over all parameters without complicated programming.
  • Microprocessor Control allows for precise liquid level control with more than 16 parameters and autofill system
  • Control panel is mounted on the top of the unit for convenient access and easy touchpad programming
  • Twenty-four tricolor LEDs continuously display actual liquid nitrogen level and high-level/low-level setpoints
  • Remote alarm contacts in-house remote alarms or connection to a Sensaphone® Telephone Dialing System
  • Keyed lock for security
Digital temperature display sensor is located under the lid. This indicates the highest temperature reading inside the tank.
The CryoPlus 1 model has 0.5 in. 45° flare, 4 ft. (1.2 m) flexible hose. The CryoPlus 2, 3 and 4 models come with 0.5 in. 45° flare, 6 ft. (1.8 m) flexible hoses.
Many optional accessories are available for these models. A complete offering of racks including vertical, square and arrowhead options to maximize storage space are available. Choose from cardboard, polycarbonate or stainlesssteel cryo boxes. The Sensaphone® Telephone Dialing System is available for added peace of mind. All accessories are Smart-Vue Remote Wireless Monitoring System Compatable: See 89496-814.
Storage capacities of up to 38,500 x 2 mL vials make the most of your laboratory storage space. Fast access to samples with large, piston-supported insulated lid.
Certifications: SFDA, cULus and CE
Accessories information: Please note the maximum number of vials that can be stored in each unit depends on the type of racks selected. The No. of vials shown below is the maximum capacity for ‘arrowhead’ racks. The No. of vials shown below for the starter kits are for VAPOR PHASE starter kits. These starter kits include square racks (89510-484) that will fit square boxes.
Ordering information: CryoPlus AutoFill Cryogenic Storage Systems are sold seperately. Optional racks, boxes, cell dividers, canisters, frames and risers allow you to create an organized/customized inventory system that provides optimum storage in liquid or vapor phase. Contact your sales representative for assistance in building the right storage configuration for your labs needs.
Caution: * Static evaporation rates are based on new container performance, no product load, styrofoam plug/lid and no lid openings. Actual working performance may vary with individual applications, ambient conditions and/or scale accuracy, excludes supply tank evaporation.