Collection Cup with Pour Spout, 6.5 oz., Globe Scientific
Supplier: Globe Scientific
5925 5920 5921 5924 5929 5926
76293-178CS 85.65 CAD
76293-178 60820-102 76293-176 89508-720 76293-174 89508-722
Collection Cup with Pour Spout, 6.5 oz., Globe Scientific
Sample Containers General Purpose Containers
This popular plastic collection cup holds up to 6.5 ounces of liquid and offers a unique pour spout to transfer liquids without spilling.
Available in crystal clear polystyrene or unbreakable polypropylene, each cup has clear graduation marks that are molded into the plastic offering measurements in both milliliters and ounces.
Ordering information: Cups are also available with attached thermometer strips or supplied with paper lids in each case.
Packaging: Cups are packaged 25 per sleeve, 20 sleeves per case (500 containers per case).