EPA Water Analysis Vials, Borosilicate Glass, Screw Thread, Without Closures, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60958A-4 60958A-6 60958A-912 60960A-4 60958A-16 60958A-11 60960A-912
66014-301CS 739.97 CAD
66014-301 66014-323 66014-345 66014-353 66009-455 66014-346 66014-355
EPA Water Analysis Vials, Borosilicate Glass, Screw Thread, Without Closures, DWK Life Sciences
Vials
Clear screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
The EPA vials are fabricated from KG-33 borosilicate glass.
Ordering information: Open top closures and PFTE-faced silicone septa or solid top PFTE-faced white rubber-lined closures are available separately.