Transfer Ribonucleic Acid (from yeast)

Supplier: MilliporeSigma

55714-1GM 55714-250MG
CA80054-304EA 1685.59 CAD
CA80054-304 CA80054-306
Transfer Ribonucleic Acid (from yeast)
Transfer Ribonucleic acid

tRNA from Baker's yeast, prepared by the method of Holley, et al.


  • A260 nm: ≥20 OD/mg in H2O; ≥16 OD/mg in buffer

MDL Number: MFCD00132200
CAS Number: 63231-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR