Protease Inhibitor Cocktail Set III, EDTA-Free, MilliporeSigma
Supplier: MilliporeSigma
539134-1ML 539134-1SET
CA80053-852EA 296.68 CAD
CA80053-852 CA80053-854
Protease Inhibitor Cocktail Set III, EDTA-Free, MilliporeSigma
Cell Culture Supplements and Additives
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases
This cocktail is recommended for use with mammalian cell and tissue extracts, but is also suitable for bacterial extracts. Supplied with a data sheet. Liquid form.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more