Perfect DNA™ Ladder, MilliporeSigma
Supplier: MilliporeSigma
70537-3 70538-3 70539-3
CA80030-884EA 370.81 CAD
CA80030-884 CA80030-886 CA80030-888
Perfect DNA™ Ladder, MilliporeSigma
Electrophoresis Reagents Molecular Weight Markers
Evenly spaced DNA markers in ready-to-load format.
  • Ready to load
  • Ideal for routine use
The Perfect DNA™ ladders contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities and cover a wide range of DNA sizes.
Delivery information: An extra vial of 6X Loading Buffer or 6X DNA Gel Loading Buffer is included.