Middlebrook 7H9 Broth Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M198-2.5KG M198-5KG
CA89224-766EA 3776.97 CAD
CA89224-766 CA89224-768
Middlebrook 7H9 Broth Base, HiMedia Laboratories
Media for Microbiology Prepared Media for Microbiology
Middlebrook 7H9 Broth Base with added enrichment is recommended for cultivation and sensitivity testing of Mycobacterium tuberculosis
Has an appearance of cream to beige homogeneous free flowing powder. Prepared Media appears as light amber colored clear solution in tubes. Store broth base below 30°C and prepared medium at 2 to 8°C.