Human TruStain FcX™, Fc Receptor Blocking Solution, BioLegend
Supplier: Biolegend
422301 422302
CA422301-BLEA 161.18 CAD
CA422301-BL CA422302-BL
Human TruStain FcX™, Fc Receptor Blocking Solution, BioLegend
Buffers Assay Buffers
Human Fc receptors (FcRs) are expressed on a variety of cells, such as monocytes, granulocytes, B cells and dendritic cells
The cells with FcR expression sometimes give false positive or false negative results of immunofluorescent staining due to the FcRs-mediated Ig Fc binding. Human TruStain FcX™ is specially formulated for blocking the FcR-involved unwanted staining without interfering with antibody-mediated specific staining of human cells. This buffer contains specialized human IgG. It is not recommended to be used for staining human IgG. Handle as biohazard agent. Soluble.