IPTG (isopropyl-β-D-thiogalactopyranoside), high purity dioxane-free, Millipore®
Supplier: MilliporeSigma
420322-10GM
420322-1GM
420322-25GM
420322-5GM
CA80056-920EA
738.33
CAD
CA80056-920
CA80056-922
CA80056-924
CA80056-926
IPTG (isopropyl-β-D-thiogalactopyranoside), high purity dioxane-free, Millipore®
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function
This preparation contains ≤0.1% dioxane by GC. White solid appearance. Protect from light. Hygrocopic.
Formula:
C₉H₁₈O₅S |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...