IntelliCAL™ MTC101 Rugged Gel Filled ORP Electrode, Hach
Supplier: Hach
MTC10110 MTC10115 MTC10130
89501-164EA 2076.8 CAD
89501-164 89501-166 89501-168 CA89501-164 CA89501-166 CA89501-168
IntelliCAL™ MTC101 Rugged Gel Filled ORP Electrode, Hach
Electrodes Redox Electrodes
IntelliCAL™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) probe with a non-refillable, gel-electrolyte reference and built-in temperature sensor
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Traceability in measurement history: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • User alerted when re-calibration is needed
  • Plug & Play: Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • These
More Product Information