Proteinase K, Tritirachium Album, MilliporeSigma
Supplier: MilliporeSigma
539480-25MG 539480-100MG
CA80017-190EA 159.35 CAD
CA80017-190 CA80017-448
Proteinase K, Tritirachium Album, MilliporeSigma
Assays Protein Assays
Serine protease that exhibits strong proteolytic activity on a wide variety of denatured and native proteins of high molecular weight
Because of its rapid proteolytic inactivation of endogenous nucleases, it can be used for isolation of mRNA and high molecular weight DNA. Preferentially cleaves bonds next to the carbonyl group of N-substituted hydrophobic, aliphatic, and aromatic amino acids. Inhibited by DFP, Hg2+, and PMSF. Not inactivated by metal chelators, sulfhydryl reagents, TLCK, or TPCK. Lyophilized from calcium acetate. Molecular Weight 28500. 50 mM Tris-HCl, 2 mM calcium acetate solubility.