Chex-All® Sterilization Tubes, Propper Manufacturing
Supplier: Propper
02600200 02600400 02600600 02600900 02601200 02600300
58753-270CS 329.62 CAD
58753-270 58753-274 58753-276 58753-278 58753-280 58753-272
Chex-All® Sterilization Tubes, Propper Manufacturing
Mailers and Shippers
Heat-sealable Chex-All® tubes are premium quality sterile packaging products for use in steam and EO.
  • Heat-Sealing Sterilization tubes allow you to be in control of the size of your pouch
  • Features triple-sealed paper/plastic film to ensure security of contents
  • Chemical indicator changes from yellow to black in steam
  • Chemical indicator changes from red to green in EO
  • Made of 2.5 mm plastic and 73 to 74 g paper
  • Triple sealed paper/plastic film provides excellent sterility protection
Chex-All tubes give you control to cut pouches to your ideal size andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable