Steri-Wrap® II, Propper Manufacturing
Supplier: Propper
31202400 31203040
89125-870CS 832.53 CAD
89125-870 76334-062 76334-064 76334-066
Steri-Wrap® II, Propper Manufacturing
Sterilization Wraps
Single use non-woven fabric sterilization wrap offers greater drapeability than standard wraps.
  • Cellulose based wrap for steam and EO sterilization
  • Efficient microbial barrier, high-level of protection against fluids and particles
  • Highly permeable meaning no wet packs
  • Variety of strength of fabric to meet your needs
Steri-Wrap® is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® II is light weight wrap designed for great drapeability.
Steri-Wrap® isMore Product Information