SP Bel-Art Round Microcentrifuge Floating Racks, Bel-Art, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18875-6401 F18875-6411
13197-152BG 116.15 CAD
13197-152 16126-544
SP Bel-Art Round Microcentrifuge Floating Racks, Bel-Art, Bel-Art Products, a part of SP
Racks Tube Racks
Molded of steam autoclavable polypropylene, these racks fit 400 mL or 1000 mL beakers.
  • Floating rack design
  • Easy to unload
  • Polypropylene construction
  • Autoclavable
Tubes remain completely immersed in a beaker while tops rest above the rack. Unload by lightly pressing down against the bench until the legs touch, releasing the tubes. Choose from 8- or 20-place racks with 10.8 mm holes for 1.5 mL microtubes. Racks are 6.4 mm (¹/₄") thick, with 19 mm (³/₄") long legs and a center handle.