SP Bel-Art Clavies™ Autoclavable Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H13185-1224 H13185-2430 H13182-1012 H13185-2436
30617-608CS 1958.45 CAD
30617-608 30617-610 490005-656 490005-658
SP Bel-Art Clavies™ Autoclavable Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Clavies® bags offer maximum protection and performance for autoclavibility.
  • Steam autoclavable at 135 °C (275 °F)
The durable 0.05 mm thick polypropylene bag is filled with used labware such as Petri dishes, membrane filters, culture plates, pipettes, and more, then autoclaved and discarded.