Fiberglass Support Rods, Ace Glass
Supplier: Ace Glass
11167-03 11167-05 11167-07 11167-09 11167-11 11167-13 11167-15 11167-32 11167-36 11167-38 11167-40 11167-42 11167-44 11167-46
89075-652EA 24.65 CAD
89075-652 89075-654 89075-656 89075-658 89075-660 89075-662 89075-664 89075-666 89075-668 89075-670 89075-672 89075-674 89075-676 89075-678
Fiberglass Support Rods, Ace Glass
Retort Rods
These Fiberglass Support Rods are used in place of aluminum rods that pit or become unsightly when used in corrosive atmosphere
Off-white or black color of these rods lends a more attractive appearance to lab setups. For use with all standard laboratory connectors.