B/T Sure™ Biological Indicators, Thermo Scientific
Supplier: Thermo Fisher Scientific
AY759X3 AY759X8 AY759X1 AY759X4 AY759X7
55710-016PK 1524.34 CAD
55710-016 58939-998 55710-014 58939-986 58939-988
B/T Sure™ Biological Indicators, Thermo Scientific
Sterilisation Indicators
Indicators verify the sterility of instruments, vials, glassware, and other products in less time than it takes to send samples to an outside lab.
  • Use to verify sterility
  • Purple color ensures sterilization
Purple color ensures sterilization. Color change from purple to yellow indicates an ineffective sterilization process. Indicators are supplied in a variety of packaging options, with each vial containing a spore population of either 105 or 106.