ACCUFORM® SSR™ Micro Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60710-1 60710-2 60710-3 60710-5 60710-10 60715-1 60715-5 60710-110 60710-310 60715-110 60715-310
66010-618CS 318.76 CAD
66010-618 66010-619 66010-620 66010-621 66010-622 66010-623 66010-624 66010-625 66010-626 66010-628 66010-629 66010-631 66010-632
ACCUFORM® SSR™ Micro Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
Storage and sample retrieval (SSR) vials are designed to provide reliable storage, retrieval, analysis, and delivery of liquid or powder samples.
These micro vials feature screw thread, open top closures, and a PTFE-faced silicone septum. These ungraduated vials are supplied assembled, and ready for use with autosamplers.