Headspace Vials, Ace Glass
Supplier: Ace Glass
5707-20 5707-22 5707-24 5707-26 5707-28 5707-30 5707-32 5707-34 5707-36 5707-40 5707-41 5707-42 5707-05 5707-06 5707-07 5707-10 5707-11 5707-12
89427-732PK 87.11 CAD
89427-732 89427-734 89427-736 89427-738 89427-740 89427-742 89427-744 89427-746 89427-748 89427-750 89427-752 89427-754 10098-814 10098-816 10098-818 10098-820 10099-042 10099-044
Headspace Vials, Ace Glass
Vials
Standard headspace vials with 20 mm finish neck for crimp seals.
Manufactured from clear, USP Type 1 borosilicate, glass. This vial has an even, flat bottom for better stirring and heating. These vials fit into Agilent or Shimadzu autosamplers.