Spherical Joints, Chemglass
Supplier: Chemglass
CG-123-01 CG-123-02
80073-164EA 190.89 CAD
80073-164 80073-166
Spherical Joints, Chemglass
Adapters Adapters, Conical Ground Joints
Special S-40 size ball and sockets for use in repairing Buchi® rotary evaporator components.
  • Size S-40 ball and sockets
  • Tooled on Duran wall tubing
Both the ball and socket are tooled on Duran 2.8mm wall tubing.