Stirrer Magnet Retrievers, Ace Glass
Supplier: Ace Glass
13672-05 13672-10
89078-010EA 61.89 CAD
89078-010 89078-012
Stirrer Magnet Retrievers, Ace Glass
Stirrers Magnetic Stirring Bars
This stirrer magnet is a 10 mm diameter pick-up rod with a powerful Alnico magnet sealed in one end and handy hang-up rip on the other.