Manifold Adapters, Ace Glass Incorporated
Supplier: Ace Glass
15429-20 15429-17
89082-832EA 380.84 CAD
89082-832 89082-834
Manifold Adapters, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This manifold adapter has a spherical socket joint to Ace-Thred.
  • Spherical socket joint
  • #7 Ace-Thred
  • 90° angle
  • With valve
Spherical 35/20 socket joint to #7 Ace-Thred, 90 degree adapter with 0-5mm valve.