Vacuum Gauge, McLeod
Supplier: Ace Glass
8728-05 8728-07 8728-09 8728-13 8728-15 8728-17 8728-41 8728-43 8728-45
89063-700EA 3879.25 CAD
89063-700 89063-702 89063-704 89063-706 89063-708 89063-710 89063-712 89063-714 89063-716
Vacuum Gauge, McLeod
Pumps Vacuum Gauges
Accurate to within ±3%
For the complete options: type B has the lowest reading at .005, type C at .010, and type D at .050.
The McLeod gauge is the most convenient and economical instrument for measuring low pressures from one micron up. The ACE McLeod gauge is offered in four ranges, so that it is possible to accurately measure any pressures between one micron and the highest reading on our D size, which is 15 mm. All gauges are fabricated from calibrated ACE Trubore glass tubing, to assure uniform and reproducible readings between gauges. This feature is also important when brokenMore Product Information