Debris Free Stirrer Bearing, Ace Glass
Supplier: Ace Glass
13445-06 13445-30 13445-09 13445-32 13445-34 13445-36 13445-46 13445-420 13445-504 13445-302 13445-426 13445-506 13445-304
89082-952EA 883.46 CAD
89082-952 89082-954 89082-958 89082-962 89082-964 89082-968 89082-970 89082-972 89082-974 89082-976 89082-978 89082-980 89082-982
Debris Free Stirrer Bearing, Ace Glass
Stirrers Stirring Tools
Vacuum tight debris free, chemically resistant stirrer bearing makes a mechanical seal against a polished glass shaft.
  • For use with 6 mm polished glass or stainless steel shafts, and not recommended for PTFE shafts
  • For use up to 500 rpm
The debris trap section consists of a PTFE sleeve for a wide range of joint and shaft diameter sizes. Bearing consists of a PTFE standard taper body with added PEEK for better stability, glass filled polypropylene screw cap, PEEK compression spring, PTFE/PEEK sleeve and a glass filled polypropylene loosening nut.