Flex-Grip Chuck, Ace Glass Incorporated
Supplier: Ace Glass
8124-26 8124-07 8124-05 8124-12 8124-25 8124-24 8124-13 8124-23 8124-10 8124-30 8124-04 8124-22 8124-20 8124-15 8124-17
89062-250EA 537.99 CAD
89062-261 89062-250 89062-252 89062-262 89062-240 89062-254 89062-242 89062-264 89062-256 89062-244 89062-258 89062-236 89062-246 89062-238 89062-248
Flex-Grip Chuck, Ace Glass Incorporated
Stirrers Stirring Tools
Nylon chuck for use with ACE stirring shafts or other shafts of the same dimensions
Chuck has flexible insert which allows for misalignment of shaft without danger of breakage.