Support Tripod, Glas-Col, Ace Glass Incorporated
Supplier: Ace Glass
12096-05 12096-10 12096-14
89076-358EA 826.36 CAD
89076-358 89076-360 89076-362
Support Tripod, Glas-Col, Ace Glass Incorporated
Supports Flask Supports
This tripod is used for 5 L, 12 L, and 22 L heating mantles with a soft glass outer covering instead of spun aluminum.
Order Now
SPECIFICATIONS
Environmentally Preferable