Friedrichs Condenser with Removable Inlet Tube, Ace Glass Incorporated
Supplier: Ace Glass
5974-02 5974-10 5974-109
89053-892PK 217 CAD
89053-892 89053-894 89053-896
Friedrichs Condenser with Removable Inlet Tube, Ace Glass Incorporated
Condensers
The top of this Friedrichs condenser is modified with #7 Ace-Thred so that the inlet tube is removable
This offers easier cleaning and greater economy if the inlet tube breaks. Fabricated from borosilicate glass. With molded spiral condensing surface, standard taper 24/40 inner joint at bottom, and standard taper reinforced outer joint at top. Jacket length is 250 mm. Hose Barb Model is for use with 3/8" I.D. tubing, size D hose connections. The #7 Ace-Thred and “Ace-Safe” hose connection on the inlet and hose barb are for use with 1/4" I.D. tubing.