Duke Standards™ Particle-Size Standards, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70889 70892 70894 70886 70887 70888 70897 70898 70896 70895 70885 70890
100496-292EA 1336.68 CAD
100496-292 100496-298 100496-302 100496-286 100496-288 100496-290 100496-308 100496-310 100496-306 100496-304 100496-284 100496-294
Duke Standards™ Particle-Size Standards, Electron Microscopy Sciences
Standards Chromatography Standards
Particle-size standards are certified for mean diameter and are traceable to the National Bureau of Standards (NBS). Duke Scientific's new line of standards in the submicrometer size range is called Nanosphere Size Standards™, and monosized polymer microspheres.
Nanosphere Size Standards are highly uniform polystyrene spheres calibrated in billionths of a meter (nanometer) with NBS traceable methodology. Nanosphere size standards are used in the calibration of electron microscopes, aerosol and liquid particle studies, chromatography columns, and laser light scattering studies. The 20 to 1000 nm range is suitable for checking the size of bacteria, viruses, ribosomes and sub-cellular components.
Monosized polymer microspheres are the most uniform spheres available. For the highest level of measurement accuracy in the 1 to 40 micron size, this series of size standards is recommended.