Bottle Top Dispenser, Wheaton, Ace Glass Incorporated
Supplier: Ace Glass
5405-22 5405-24 5405-41 5405-42
89051-646EA 1997.17 CAD
89051-646 89051-648 89051-650 89051-652
Bottle Top Dispenser, Wheaton, Ace Glass Incorporated
Bottle Top Dispensers
These unbreakable Bottle Top Dispensers are designed for use with glass bottles with a 33mm screw neck finish.
Rotate handpiece and receiver to adjust to desired volume (increments of 0.1mL). To dispense, raise handpiece fully, then depress. Designed for 33mm screw necks, although adapters for other size necks are available.