Flat Flange Reaction Head, 4 Neck, Ace Glass
Supplier: Ace Glass
6515-46 6515-50 6515-54 6515-58 6515-47 6515-48 6515-60 6515-62 6515-64
89055-098EA 1034.53 CAD
89055-098 89055-100 89055-102 89055-104 76106-996 76106-998 76107-000 76107-002 76107-004 76107-006
Flat Flange Reaction Head, 4 Neck, Ace Glass
Lids Reaction Vessel Lids
For use with ground flat flange flasks
With three in-line standard taper joints and a standard taper 10/30 side joint for thermometer. Fabricated from borosilicate glass.